missing translation for 'onlineSavingsMsg'
Learn More
Yttrium pieces, 99.9% (REO), Thermo Scientific Chemicals Product Code.: 11336597

Yttrium pieces, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11336597
50 g, Glass bottle; Ampouled under argon
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle; Ampouled under argon
Under Argon
This item is not returnable. View return policy

Product Code. 11336597

Brand: Thermo Scientific Alfa Aesar 000615.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Quantity 50 g
Percent Purity 99.9%
Packaging Glass bottle; Ampouled under argon
Chemical Name or Material Yttrium pieces

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ24K021 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number000615.18,000615.09
Lot NumberS14K019 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number000615.18,000615.09
Lot NumberZ09I033 Certificate TypeCertificate of Analysis Date06/01/2023 Catalog Number000615.18,000615.09
Lot NumberQ04I044 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000615.18,000615.09
Lot NumberU13G030 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number000615.18,000615.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium pieces, 99.9% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.