missing translation for 'onlineSavingsMsg'
Learn More
Yttrium powder, -40 mesh, 99.6% (REO), Thermo Scientific Chemicals Product Code.: 11339648

Yttrium powder, -40 mesh, 99.6% (REO), Thermo Scientific Chemicals

Product Code. 11339648
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11339648

Brand: Thermo Scientific Alfa Aesar 044147.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Color Gray
Appearance Gray powder
Assay Percent Range 99.6%
Quantity 5 g
Mesh Size -40 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Yttrium, Rare Earth Oxides Content

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX30K025 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number044147.14,044147.06
Lot NumberT11I019 Certificate TypeCertificate of Analysis Date22/07/2022 Catalog Number044147.14,044161.AE,044147.06
Lot NumberK19N21 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044147.14,044161.AE,044147.06
Lot NumberR13H035 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044147.14,044161.AE,044147.06
Lot Number61401099 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number044147.14,044161.AE,044147.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Yttrium powder, -40 mesh, 99.6% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.