missing translation for 'onlineSavingsMsg'
Learn More

Yttrium solution for ICP/MS 100mg/L HNO3 matrix, Fisher Chemical™

Product Code. 12967313
Click to view available options
Quantity:
100 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Concentration 100mg/L
CAS 7697-37-2
Packaging Bottle
Quantity 100 mL
Molecular Formula HNO3
MDL Number MFCD00011349
InChI Key GRYLNZFGIOXLOG-UHFFFAOYSA-N
SMILES O[N+]([O-])=O
IUPAC Name nitric acid
Molecular Weight (g/mol) 63.01
PubChem CID 944
ChEBI CHEBI:48107
Solvent or Matrix HNO3
Chemical Name or Material Yttrium
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'