missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Standard For ICP, 2% HNO3 in 1000μg/mL, Assurance Grade, SPEX CertiPrep™

Product Code. 10102682
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Y1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-65-5, 7697-37-2, 7732-18-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Packaging Triple-leached LDPE bottle
CAS Min % 1.0, 5.0, 94.0
Quantity 500 mL
CAS Max % 1.0, 5.0, 94.0
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLY2-2Y, -2T, -2X    10,000    PLY2-3Y, -3X
Solvent or Matrix 2% HNO3
Chemical Name or Material Yttrium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000 ug/mL Yttrium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : nitric acid

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'