Learn More
Tocris Bioscience™ Z-DEVD-FMK Caspase-3 Inhibitor
Caspase-3 Inhibitor
Brand: Tocris Bioscience™ 8009/1
240.17 GBP valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
Z-DEVD-FMK Caspase-3 Inhibitor is a cell-permeable, irreversible inhibitor of caspase-3/CPP32. It inhibits tumor cell apoptosis. In vivo, it has a neuroprotective effect in rat hippocampus following seizures. It also significantly reduces post-traumatic apoptosis, and improved neurological recovery, before and after induced traumatic brain injury in rat.Specifications
634911-78-7 | |
Caspase-3 | |
0.95 |
1 mg | |
C30H41FN4O12 |