missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 16 Peptide
Z'-LYTE™ Kinase Assay Kit - Ser/Thr 16 Peptide
Brand: Invitrogen™ PV3802
Code : 50
Additional Details : Weight : 0.09000kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon CDC25C, containing Ser-198.Specifications
Z´-LYTE™ | |
FRET | |
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
Methylcoumarin | |
800 x 20 μL Assays | |
CDC25C residues 193-204, centered at Ser-198 (ELMEFSLKDQEA) | |
800 Reactions | |
End Point | |
Peptide-Based Substrate | |
ACVR1B, ADRBK1, ADRBK2, GRK4, GRK5, GRK6, GRK7, PLK1, PLK2, PLK3 |
Serine/Threonine Kinases | |
Serine/Threonine Kinase Assay | |
Biochemical phospho-specific activity (no antibody) | |
Kinase Assay Kit | |
384-well plate | |
405⁄465, 535 | |
JBC 278:25277 (2003) | |
Dry Ice | |
GRK7, ADRBK1, ADRBK2, ACVR1B, PLK2, PLK3, GRK6, GRK5, GRK4, PLK1 |
For Research Use Only. Not for use in diagnostic procedures.