missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 18 Peptide
Z'-LYTE Kinase Assay Kit - Ser/Thr 18 Peptide
Marque: Invitrogen™ PV4319
Code : 50
Informations supplémentaires : Weight : 0.50000kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon an optimized synthetic peptide substrate for DYRK1A, and containing an R-P-X-(S⁄T)-P consensus sequence, where X is any residue.Spécification
Z´-LYTE™ | |
FRET | |
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
Methylcoumarin | |
800 x 20 μL Assays | |
RRFRPASPLRGPP | |
800 Reactions | |
End Point | |
Peptide-Based Substrate | |
CLK3, DYRK1A, DYRK1B, HIPK4, SRPK1, STK23 |
Serine/Threonine Kinases | |
Serine/Threonine Kinase Assay | |
Biochemical phospho-specific activity (no antibody) | |
Kinase Assay Kit | |
384-well plate | |
405⁄465, 535 | |
JBC 275, pg 2431 (2000) | |
Dry Ice | |
DYRK1A, HIPK4, SRPK1, DYRK1B, CLK3, STK23 |
For Research Use Only. Not for use in diagnostic procedures.