Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 20 Peptide
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.
Brand: Thermo Scientific™ PV4532
Code : 50
Additional Details : Weight : 0.09000kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon a synthetic peptide substrate for NDR1.
Specifications
Z ́-LYTE™ | |
Biochemical phospho-specific activity (no antibody) | |
405/465, 535 | |
FRET | |
800 Reactions | |
End Point | |
Dry Ice | |
Peptide-Based Substrate | |
Serine/Threonine Kinases |
Serine/Threonine Kinase Assay | |
Z ́-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
384-well Plate | |
Methylcoumarin | |
800 x 20 μL | |
EMBO J. vol. 17, pp. 5913-22 (1998) | |
Based on Synthetic Peptide Substrate for NDR1 (KKRNRRLSVA) | |
PAK4, RPS6KA6, PAK7, PAK6, PAK2, PAK3 | |
PAK2, PAK3, PAK4, PAK6, PAK7, RPS6KA6 |
For Research Use Only. Not for use in diagnostic procedures.