missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 7 Peptide
Z'-LYTE™ Kinase Assay Kit - Ser/Thr 7 Peptide
Brand: Invitrogen™ PV3180
Code : 50
Additional Details : Weight : 3.18000kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide contains a PKC consensus sequence (R⁄K)1-3(X)0-2 (S⁄T) (X)0-2(R⁄K)1-3, where X is any residue.Specifications
Z´-LYTE™ | |
FRET | |
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
Methylcoumarin | |
800 x 20 μL Assays | |
PKC consensus sequence (R⁄K)1-3(X)0-2 S⁄T (X)0-2(R⁄K)1-3 | |
800 Reactions | |
Dry Ice | |
MAP4K5, IRAK4, PRKCQ, NEK9, PRKCD, MAP3K9, PASK, PIM2, NEK2, PRKCG, SRPK2, STK4, NEK1, MAP4K2, MINK1, CHEK2, PIM1, PKN1, PRKCA, PRKCI, TAOK2, MAP4K4, RPS6KB1, PHKG1, PRKCZ, ROCK1, PHKG2, PRKCB1, STK24, STK3, PRKCB2, STK25, PRKCE, PRKCH, MST4 |
Serine/Threonine Kinases | |
Serine/Threonine Kinase Assay | |
Biochemical phospho-specific activity (no antibody) | |
Kinase Assay Kit | |
384-well plate | |
405⁄465, 535 | |
End Point | |
Peptide-Based Substrate | |
ROCK1, RPS6KB1, SRPK2, STK3, STK4, STK24, STK25, TAOK2, NEK2, NEK9, PASK, PHKG1, PHKG2, PIM1, PIM2, PKN1, PKC, CHEK2, IRAK4, MAP3K9, MAP4K2, MAP4K4, MAP4K5, MINK1, MST4, NEK1 |
For Research Use Only. Not for use in diagnostic procedures.