missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Ser/Thr 9 Peptide
Z'-LYTE Kinase Assay Kit - Ser/Thr 9 Peptide
Brand: Invitrogen™ PV3324
Code : 50
Additional Details : Weight : 0.00900kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon glycogen synthase I, containing Ser-641.Specifications
Z´-LYTE™ | |
FRET | |
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
Methylcoumarin | |
800 x 20 μL Assays | |
glycogen synthase I (PRPASVPP(pS)P(pS)) | |
800 Reactions | |
End Point | |
Peptide-Based Substrate | |
CLK1, DYRK3, DYRK4, GSK3A, GSK3B, HIPK1 |
Serine/Threonine Kinases | |
Serine/Threonine Kinase Assay | |
Biochemical phospho-specific activity (no antibody) | |
Kinase Assay Kit | |
384-well plate | |
405⁄465, 535 | |
JBC, 279, pp.2490-2498 (2004) | |
Dry Ice | |
GSK3A, GSK3B, HIPK1, DYRK4, CLK1, DYRK3 |
For Research Use Only. Not for use in diagnostic procedures.