missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 1 Peptide
Z'-LYTE™ Kinase Assay Kit - Tyrosine 1 Peptide
Brand: Invitrogen™ PV3190
Code : 50
Additional Details : Weight : 0.09000kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon an optimal peptide substrate for SRC.Specifications
Z´-LYTE™ | |
FRET | |
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions. | |
Methylcoumarin | |
800 x 20 μL Assays | |
Src Substrate (EEEIYGEFE) | |
800 Reactions | |
End Point | |
Peptide-Based Substrate | |
BLK1, BMX, BTK, CSF1R, EPHA3, EPHA5, EPHB4, ERBB4, FES, FRK, IGF1R, ITK, KDR, MATK, NTRK1, NTRK2, NTRK3, PTK2B, PTK6, ROS1, SRMS |
Tyrosine Kinases | |
Tyrosine Kinase Assay | |
Biochemical phospho-specific activity (no antibody) | |
Kinase Assay Kit | |
384-well plate | |
405⁄465, 535 | |
Nature, 373, pp. 536-9 (1995) | |
Dry Ice | |
CSF1R, NTRK2, PTK6, IGF1R, MATK, NTRK3, ROS1, BMX, PTK2B, EPHA3, EPHB4, ERBB4, KDR, NTRK1, SRMS, FRK, ITK, BTK, FES, BLK1, EPHA5 |
For Research Use Only. Not for use in diagnostic procedures.