Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 2 Peptide
Z'-LYTE Kinase Assay Kit - Tyrosine 2 Peptide
Brand:  Invitrogen™ PV3191
Product Code. 12080176
  • £562.00 / 800 reactions
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon an optimal peptide substrate for ABL.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Z´-LYTE™
FRET
Z´-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
Methylcoumarin
800 x 20 μL Assays
Abl Substrate (EAIYAAPF)
800 Reactions
End Point
Peptide-Based Substrate
ABL1, ABL2, CSK2, EPHA1, EPHA4, EPHA8, EPHB1, EPHB2, MERTK, MET, RET, SRC, SRCN1, SYK, TYRO3, YES1, EPHB3, FGR, FLT, FYN, HCK, LCK, LYNA, LYNB
Tyrosine Kinases
Tyrosine Kinase Assay
Biochemical phospho-specific activity (no antibody)
Kinase Assay Kit
384-well plate
405⁄465, 535
Nature, 373, pp 536-9 (1995)
Dry Ice
ABL1 G250E, ABL1 Y253F, RET Y791F, SYK, TYRO3, EPHB2, LYNA, MERTK, RET V804L, SRCN1, ABL1 E255K, EPHA4, FYN, RET, ABL1 T315I, FLT3, LCK, YES1, MET, ABL1, ABL2, FGR, HCK, EPHA1, EPHA8, EPHB3, LYNB, EPHB1, SRC, CSK