missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 3 Peptide
Z'-LYTE™ Kinase Assay Kit - Tyrosine 3 Peptide
Brand: Invitrogen™ PV3192
Code : 50
Additional Details : Weight : 0.09000kg
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon MAPK14, containing Tyr-182.Specifications
Z'-LYTE™ Tyrosine 3 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Liquid, Tyrosine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis | |
Protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acute-lyte and kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity | |
800 x 20μL | |
Z'-LYTE™ Tyrosine 3 Peptide Kinase Assay Kit | |
384-well plate |
Liquid | |
Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer | |
–80°C | |
800 x 20μL |
For Research Use Only. Not for use in diagnostic procedures.