Learn More
Thermo Scientific™ Z'-LYTE™ Kinase Assay Kit - Tyrosine 7 Peptide
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format.
Brand:  Thermo Scientific™ PV4305
Product Code. 10492902
  • £548.00 / Pack of 800
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
A coumarin and fluorescein-labeled peptide substrate which can be used for kinase screening in an antibody-free format. This peptide is based upon LAT, containing Tyr-200.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Z'-LYTE™ Tyrosine 7 Peptide Kinase Assay Kit, 800 x 20μL, 800rxns, Liquid, Tyrosine Kinase Assay Type, Peptide-Based Substrate, 535nm Emission, 405/465nm Excitation, Methylcoumarin Label, End Point Readout, Threonine Kinases Target Enzyme, FRET Technique, Used for Pharma and Biopharma, Protein Assays and Analysis
Liquid
Z'-LYTE™ Ser/Thr 11 peptide, Z'-LYTE™ Ser/Thr 11 phospho-peptide, 5X kinase buffer A, ATP, development reagent B, development buffer
Tyrosine Kinase Assay
Z'-LYTE™ Kinase Assay Kits contain all necessary reagents for performing primary or secondary screens. See COA for specific storage instructions Most kit components should be stored at -20°C, and some kit components should be stored at -80°C. See COA for specific storage instructions.
800 x 20μL, 800 x 20 μL
405/465, 535 nm
800 Reactions
Biochem. J. 356 p 461-71 (2001)
Based on LAT, Residues 195 to 208, Centering Y-200 (ESIDDYVNVPESGES)
ZAP70
ZAP70
Z ́-LYTE™
Pharma and biopharma, protein biology, drug discovery and development, target and lead identification and validation, kinase biology, biochemical kinase assays, z and acutelyte™ kinase assays, enzyme and protein activity assays, protein assays and analysis, kinase activity, FRET
800 x 20μL
Biochemical phospho-specific activity (no antibody)
Z'-LYTE™ Tyrosine 7 Peptide Kinase Assay Kit
384-well plate
Methylcoumarin
End Point
Dry Ice
Peptide-Based Substrate
Tyrosine Kinases