missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZACN (extracellular) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577765
Additional Details : Weight : 0.01000kg
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 μL of deionized water.
ZACN is a zinc-activated ligand-gated ion channel that defines a new subgroup of the cysteine-loop superfamily of ligand-gated ion channels.Specifications
ZACN (extracellular) | |
Polyclonal | |
Unconjugated | |
ZACN | |
ZACN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
353174 | |
-20°C | |
Lyophilized |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.8 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
Q401N2 | |
ZACN | |
Peptide CNFELLHFPRDHSN, corresponding to amino acid residues 157-170 of human zinc-activated channel (ZAC) | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |