missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zacopride hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Highly potent 5-HT3 receptor antagonist (Ki = 0.38 nM) and 5-HT4 receptor agonist (Ki = 373 nM). Antiemetic and anxiolytic following systemic administration in vivo.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 346.26 |
Percent Purity | >99% |
Chemical Name or Material | Zacopride hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'