missing translation for 'onlineSavingsMsg'
Learn More
Zanamivir monohydrate, Thermo Scientific Chemicals Product Code.: 16554364

Zanamivir monohydrate, Thermo Scientific Chemicals

Product Code. 16554364
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 16554364

Brand: Thermo Scientific Acros 462960010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector ZANAMIVIR
CAS 1260601-68-0
Melting Point 256°C
Color White
Loss on Drying 4 % to 9 % (1 g, 105°C) (vacuum)
Infrared Spectrum Conforms
Packaging glass bottle
Molecular Formula C12 H20 N4 O7 . H2 O
MDL Number MFCD00866966
Quantity 1 g
Specific Rotation 36° to 38° (20°C, 589 nm) (1 % aq. soln.)(20°C, 589 nm) (on dry substance)
Molecular Weight (g/mol) 350.33
Formula Weight 350.33
Physical Form Crystalline Powder
Chemical Name or Material Zanamivir monohydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0449078 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog Number462960010
Lot NumberA0445791 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number462960010
Lot NumberA0444825 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number462960010
Lot NumberA0411117 Certificate TypeCertificate of Analysis Date08/11/2021 Catalog Number462960010

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zanamivir monohydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.