Learn More
Labcon™ ZAP™ 1200 μL Aerosol Filter Pipette Tips for Sartorius™ Pipettes
These 1250 μL aerosol filter pipette tips have UltraFine™ points and come in racks.
Brand:  Labcon™ 1194-965-006-9
Additional Details : Weight : 1.56200kg
Product Code. 17547876
  • £77.60 / Pack of 576
Estimated Shipment: 02-09-2024
to see stock.
Add to basket
Description
Description
  • Fits Sartorius™ 1200 μL Pipettes
  • Protease, Human DNA, ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
  • Validated Sterilization (ISO 11137) for use in Research and Medical labs
  • Made in USA
Specifications
Specifications
1200 μL
Sterile
Purple
88.9 mm
Protease, Human DNA, ATP, RNase/DNase, Endotoxin-free
E-beam
Rectangle
Single-use
No
Filtered
Clear
Autoclavable
ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Micropipetting
Rack
96 Tips/Rack, 6 Racks/Pk., 8 Pk./Cs.
Sartorius
Polypropylene Resin