missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zardaverine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Phosphodiesterase inhibitor, selective for PDE3 and 4 (IC50 values are 0.5 and 0.8 μM respectively).
Specifications
Quantity | 50 mg |
Formula Weight | 268.22 |
Percent Purity | >98% |
Chemical Name or Material | Zardaverine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'