missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL

Perform high-throughput protein recovery (7K MWCO) and sample desalting for mass spectrometry, HPLC, capillary electrophoresis, metabolite screening, and assay development with ready-to-use Thermo Scientific Pierce Zeba desalting chromatography columns, plates, and cartridges.

Brand:  Thermo Scientific™ 89882

128.25 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 10341164

  • £137.00 / Pack of 25

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Spin Desalting Column
Spin Column
Size-exclusion, Proprietary Resin
Zeba™
0.5 mL
Upon receipt store at 4°C.
Buffer Exchange, Protein
7.0 kDa
25 Columns
30 to 130 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Zeba™ Spin Desalting Columns, Plates, and Cartridges, 7K MWCO, 0.5–100 mL > Zeba™ Spin Desalting Columns, 7K MWCO, 0.5 mL; 25 Columns

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.