missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebularine, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Orally active DNA methyltransferase inhibitor. Inhibits tumor cell proliferation (IC50 = 120 μM) and re-activates silenced genes in T24 bladder carcinoma cells. Inhibits cytidine deaminase (Ki ∽ 2 μM); induces differentiation of mesenchymal stem cells into cardiomyocytes.
Specifications
Specifications
CAS | 3724-10-5 |
Molecular Formula | C8H8O2S |
Quantity | 10 mg |
Synonym | 2-methylthio benzoic acid, benzoic acid, 2-methylthio, o-methylthio benzoic acid, benzoic acid, o-methylthio, 2-methylsulfanyl benzoic acid, 2-methylmercaptobenzoic acid, 2-carboxyphenyl methyl sulfide, acide methyl-s-2-benzoique, 2-methylmercapto benzoic acid, acide methyl-s-2-benzoique french |
InChI Key | LWJQGKJCZOGGPJ-UHFFFAOYSA-N |
SMILES | CSC1=CC=CC=C1C(=O)O |
IUPAC Name | 2-methylsulfanylbenzoic acid |
Molecular Weight (g/mol) | 168.21 |
PubChem CID | 19498 |
Formula Weight | 228.2 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction