missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zenon™ Mouse IgG1 Labeling Kits

Product Code. 10482292
Click to view available options
Quantity:
10 Reactions
25 Reactions
25 Reactions kit
3 x 10 μL
50 Reactions
50 Reactions kit
Label or Dye:
APC (Allophycocyanin)
Alexa Fluor 350
Alexa Fluor 405
Alexa Fluor 488
Alexa Fluor 488, Alexa Fluor 555, Alexa Fluor 647
Alexa Fluor 546
Alexa Fluor 555
Alexa Fluor 568
Alexa Fluor 594
Alexa Fluor 647
Alexa Fluor 647-R-Phycoerythrin
Alexa Fluor 700
Biotin
HRP (Horseradish Peroxidase)
Pacific Blue
R-PE (R-Phycoerythrin)
Unit Size:
1 set
Each
Excitation/Emission:
346/442 nm
402/421 nm
410/455 nm
495, 555, 650/519, 565, 668 nm
496, 546, 565/578 nm
496/519 nm
496/668 nm
555/565 nm
556/573 nm
578/603 nm
590/617 nm
650/660 nm
650/668 nm
696/719 nm
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10482292

Brand: Invitrogen™ Z25054

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

Contains 1 vial of Zenon HRP mouse IgG1 labeling reagent (125 μL), and 1 vial of Zenon blocking reagent (mouse IgG, 125 μL).

Simplify your multicolor staining and flow cytometry experiments with our Zenon™ mouse IgG1 labeling kits.

Simplify your multicolor staining and flow cytometry experiments with our Zenon™ mouse IgG1 labeling kits. These antibody labeling kits are ideal for flow cytometry (e.g., FACS) experiments because they enable the simultaneous labeling of different target cells and tissues with differently labeled mouse monoclonal antibodies in a single staining protocol. Multiple Zenon™ antibody complexes may be prepared individually and used in a multicolor stain after using the Zenon™ blocking reagent.

Achieve fast, versatile, and reliable fluorophore-, biotin-, or enzyme-labeled primary antibodies with the Zenon™ mouse IgG1 labeling kits. These kits utilize Alexa Fluor fluorophores, biotin, Pacific Blue, or enzymes such as R-phycoerythrin and allophycocyanin, which are attached to monovalent, affinity purified Fab fragments. The Fab fragments, in turn, are directed against and bind with the Fc portion of IgG1 primary antibodies. Only a small amount of starting material is required, and the method is optimized for efficient labeling of antibodies in serum, ascites fluid, or hybridoma suspensions. Because the Zenon labeling method is based on immunoselectivity, it does not require the removal of exogenous proteins (such as serum) or amine-containing buffers from the target antibody, simplifying the process.

Zenon labeling technology greatly simplifies the use of multiple mouse-derived antibodies in the same staining protocol. Zenon tricolor labeling kits contain sufficient materials for 10 labeling reactions of each of three different fluorescent colors.

Important features of Zenon labeling technology:
• Labeled antibodies are typically ready to use in 10 minutes
• Requires only 1–20 μg primary antibody
• Simple—no purification required
• Flexible—choose from different fluorophores, biotin, HRP, alkaline phosphatase
• Multiplex with other mouse monoclonal antibodies simultaneously
• Can be used in a variety of applications including ICC, IHC, flow cytometry, and cell imaging.

Advantages of using Zenon antibody labeling kits:

Cost savings
Zenon antibody labeling kits offer a cost-conscious and reproducible method of tagging as little as 0.4 μg in 2 μL of primary antibody, with minimal waste of expensive or difficult-to-obtain antibodies, or excessive washing steps that pose the risk of product loss.

Sensitivity
Label your primary antibodies without compromising their antigen binding affinity: Zenon dye- and enzyme-labeled Fab fragments, which are targeted to the Fc tail, are affinity purified during their preparation to ensure high affinity and selectivity for the Fc portion of the corresponding primary antibody. The procedure for chemical labeling of the Zenon Fab fragments protects their Fc-binding site, resulting in more active labeling reagents.

Speed
No purification procedure is required prior to using Zenon Fab fragments in your laboratory applications. Formation of the Fab-antibody complex occurs in fewer than five minutes, followed by a five-minute blocking step. During this time, almost all the primary antibody in the mixture is labeled with the labeled Fab fragments.

Simplicity
The Fab-antibody complexes display fluorescence or enzymatic activity that is similar in intensity to that of directly labeled primary antibodies. Varying the extent of the antibody labeling is as simple as changing the amount of added Zenon labeling reagent during the reaction. Once the labeling complexes are formed, they can used immediately, without need for antibody purification.

Reliability
The Zenon Fab-antibody complex is stable and allows subsequent or simultaneous labeling of different target cells and tissues with different complexes. After staining, an aldehyde-based fixing step may be used to prevent the transfer of different labels between different primary antibodies, preserving the initial staining pattern.

Customization
We offer custom antibody conjugation services that are efficient and confidential, and we stand by the quality of our work. We are ISO 13485:2000 certified.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Labeling Kit
Quantity 25 Reactions
Content And Storage Contains 1 vial of Zenon HRP mouse IgG1 labeling reagent (125 μL), and 1 vial of Zenon blocking reagent (mouse IgG, 125 μL).

Store in refrigerator (2–8°C) and protect from light.
Labeling Scale < 1–20 μg
Product Line Zenon
Label or Dye HRP (Horseradish Peroxidase)
Color Colorless
Labeling Target IgG1
Species Mouse
Label Type Enzyme Labeled

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.