missing translation for 'onlineSavingsMsg'
Learn More
Zeolite beta, hydrogen, Thermo Scientific Chemicals Product Code.: 15452507

Zeolite beta, hydrogen, Thermo Scientific Chemicals

Product Code. 15452507
500 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 15452507

Brand: Thermo Scientific Alfa Aesar 045875.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)Zeolite β, hydrogen is used as a molecular sieve, filter, adsorbent, catalyst, drying agent, cation exchanger, dispersing agent and detergent builder. It serves as an acid catalyst and utilized as an alternative to the known methods for protection of alcohols as tetrahydropyranyl ethers and the deprotection of tetrahydropyranyl ethers. It is also used in the stereoselective Meerwein-Ponndorf-Verley reduction of ketones. Further, it is used as an efficient and recyclable catalyst for the tetrahydropyranylation of alcohols and phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Zeolite beta is used in isomerization of waxes, and Friedel Crafts reactions (alkylation and acylation)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1318-02-1
Molecular Formula Al2O5Si
Molecular Weight (g/mol) 162.043
MDL Number MFCD00132601
InChI Key HNPSIPDUKPIQMN-UHFFFAOYSA-N
Synonym kaolinite, montmorillonite aluminum pillared clay, silica-alumina catalyst support, grade 135, aidplusocma, bactekillermb, fiberfrax™, bactekillerbm101a, bactekillerbm102a, bactekillerbm501a, kaolinite, naturalShow MoreShow Less
PubChem CID 9942228
IUPAC Name dioxosilane;oxo(oxoalumanyloxy)alumane
SMILES O=[Al]O[Al]=O.O=[Si]=O

Specifications

Color White
Physical Form Powder
Quantity 500 g
Solubility Information Insoluble in water.
Chemical Name or Material Zeolite beta, hydrogen

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX25K027 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog Number045875.22,045875.14,045875.36
Lot NumberS14K040 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number045875.22,045875.14,045875.36
Lot NumberM09K023 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number045875.22,045875.14,045875.36
Lot NumberM17K011 Certificate TypeCertificate of Analysis Date25/01/2024 Catalog Number045875.22,045875.14,045875.36
Lot NumberS26J030 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog Number045875.22,045875.14,045875.36,045875

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zeolite beta, hydrogen, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.