missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFYVE21 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24806-1-AP-20UL
This item is not returnable.
View return policy
Description
ZF21 was initially identified as protein that could bind to the cytoplasmic tail of MT1-MMP (Membrane-type 1 matrix metalloproteinase), a potent invasion-promoting protease. ZF21 is a member of a protein family characterized by the presence of a phosphatidylinositol 3-phosphate-binding FYVE domain and regulates focal adhesions (FAs) and cell movement. Knockdown of ZF21 expression resulted in a delay of FA disassembly following induction of synchronous disassembly of FAs by nocodazole treatment, suggesting that ZF21 is involved in FA disassembly. ZF21 contains a noncanonical pleckstrin homology domain that is a possible therapeutic target to treat metastatic cancer.Specifications
ZFYVE21 | |
Polyclonal | |
Unconjugated | |
ZFYVE21 | |
ZFYVE21 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79038 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9BQ24 | |
ZFYVE21 | |
ZFYVE21 Fusion Protein Ag20024 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |