missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc 2,4-pentanedionate monohydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
Zinc Metal acetylacetonates are used as catalysts for polymerization of olefins and transesterification. It is used as a a stabilizer PVC.They are used as solvents, lubricant additives, paint drier, and pesticides. They are used in glass coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 136°C to 138°C (decomposition) |
Density | 1.6 g/mL |
Quantity | 50 g |
Solubility Information | Soluble in alcohol; Slightly soluble in water |
Formula Weight | 281.60 (263.59 Anhydrous) |
Physical Form | Crystalline |
Chemical Name or Material | Zinc 2,4-pentanedionate monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'