Learn More
Zinc acetate, 99%, pure, Thermo Scientific Chemicals
£52.36 - £149.75
Chemical Identifiers
CAS | 557-34-6 |
---|---|
Molecular Formula | C4H6O4Zn |
Molecular Weight (g/mol) | 183.48 |
MDL Number | MFCD00012454 |
InChI Key | DJWUNCQRNNEAKC-UHFFFAOYSA-L |
Synonym | zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4 |
PubChem CID | 11192 |
ChEBI | CHEBI:62984 |
IUPAC Name | zinc;diacetate |
SMILES | CC(=O)[O-].CC(=O)[O-].[Zn+2] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10552062
|
Thermo Scientific Acros
370080250 |
25 g | Glass bottle |
£52.36
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10292812
|
Thermo Scientific Acros
370081000 |
100 g | Glass bottle |
£149.75
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
557-34-6 | |
183.48 | |
DJWUNCQRNNEAKC-UHFFFAOYSA-L | |
11192 | |
zinc;diacetate |
C4H6O4Zn | |
MFCD00012454 | |
zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4 | |
CHEBI:62984 | |
CC(=O)[O-].CC(=O)[O-].[Zn+2] |
Specifications
83°C to 86°C | |
Gray-Green to Yellow | |
98.5% min. (Complexometry) | |
(CH3CO2)2Zn | |
15, 10327 | |
DJWUNCQRNNEAKC-UHFFFAOYSA-L | |
zinc;diacetate | |
11192 | |
183.48 | |
Pure | |
1% max. |
557-34-6 | |
Powder | |
C4H6O4Zn | |
MFCD00012454 | |
zinc acetate, zinc diacetate, zinc ii acetate, acetic acid, zinc salt, dicarbomethoxyzinc, acetic acid, zinc ii salt, zinc di acetate, galzin, zinc acetate anhydrous, siltex cl 4 | |
CC(=O)[O-].CC(=O)[O-].[Zn+2] | |
183.48 | |
CHEBI:62984 | |
99% | |
Authentic | |
Zinc acetate |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiti
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-170-2
- Zinc acetate
Signal Word
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H400-Very toxic to aquatic life.
- H319-Causes serious eye irritation.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only