missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc acetate dihydrate, 98+%, ACS reagent
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
10kg
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 237.0°C |
| Color | White |
| pH | 6.0 to 7.0 (5% soln. at 25°C) |
| Physical Form | Crystals |
| Quantity | 500 g |
| Assay Percent Range | 98.0 to 101% (Complexometry) |
| Linear Formula | (CH3CO2)2Zn·2H2O |
| Merck Index | 15,10327 |
| Solubility Information | Solubility in water: 430g/L (20°C). Other solubilities: soluble in alcohol |
| Formula Weight | 219.5 |
| Show More |
Safety and Handling
Product Identifier
- Zinc acetate dihydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H411-Toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Berichtigung von Produktinhalten
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu