missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Classic Vortex Mixer

Combines the highest performance ratings in terms of speedwith excellent reliability and safety. Fisherbrand™ Classic Vortex Mixer includes two operating modes, the possibility to change the vibration frequency, and a wide range of accessories.

Brand:  Fisherbrand™ F202A0173FI

187.62 GBP valid until 2024-12-31
Use promo code "21745" to get your promotional price.


Product Code. 11726744

  • £260.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Combines the highest performance ratings in terms of speedwith excellent reliability and safety. Fisherbrand™ Classic Vortex Mixer includes two operating modes, the possibility to change the vibration frequency, and a wide range of accessories.

Original Design
  • Compact in height and area
  • Stability is optimized with selection lever related to operating mode
  • 3 anti-slip rubber feet for staying in required position
Multi-Environment Operation
  • Variable speed operation, electronically controlled up to 3000 rpm
  • Large choice of accessories to suit tubes, plates and flasks
  • Universal voltage / frequency model
Designed to Last
  • Accessories fit by simple pressure
  • Optimum resistance to chemicals from technopolymer casing
  • Excellent stability with zinc alloy base
  • IP 42 protection against particles and liquids
Specifications

Specifications

4.5 mm
15 W
50/60 Hz
220 mm
2.75 in.
7.1 in.
8.7 in.
2.7 kg
IP 42
0 to 3000 rpm
Vortex Mixer
Zinc Alloy Base, Technopolymer Platform
7.1 x 8.7 x 2.8 in. (180 x 220 x 70 mm)
70 mm
180 mm
220 mm
Classic
Touch, Continuous
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.