Learn More
Fisherbrand™ Zinc Alloy Bossheads
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Zinc Alloy Bossheads come with plated clamping screws with plastic heads.
Brand:  Fisherbrand™ 9506/P
Product Code. 10117184
  • £123.17 / Pack of 5
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
For securely keeping clamps in place on laboratory scaffolding. Fisherbrand™ Zinc Alloy Bossheads come with plated clamping screws with plastic heads.
  • For rods up to 16mm in diameter
  • Supplied in packs of 5
Specifications
Specifications
5/Pk.
Zinc Alloy
For use with rod up to 16mm diameter
Bosshead Clamp
Plastic headed clamping screws
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Zinc Alloy Bossheads > Includes: Plastic headed clamping screws