Learn More
Zinc bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
£25.43 - £270.35
Chemical Identifiers
CAS | 168106-25-0 |
---|---|
Molecular Formula | C4F12N2O8S4Zn |
Molecular Weight (g/mol) | 625.651 |
MDL Number | MFCD16621474 |
InChI Key | QEORIOGPVTWFMH-UHFFFAOYSA-N |
Synonym | Zinc triflimide; Zinc trifluoromethanesulfonimide |
PubChem CID | 131849436 |
IUPAC Name | zinc;bis(trifluoromethylsulfonyl)azanide |
SMILES | C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15482787
|
Thermo Scientific Alfa Aesar
046880.MD |
250 mg |
£25.43
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15462787
|
Thermo Scientific Alfa Aesar
046880.03 |
1 g |
£70.25
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472787
|
Thermo Scientific Alfa Aesar
046880.06 |
5 g |
£270.35
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zinc bis(trifluoromethylsulfonyl)imide is used as a Catalyst for Enantioselective Friedel-Crafts alkylation of pyrroles with β-CF3 acrylates, Stereoselective Diels-Alder reaction, Tandem cyclopropane ring-opening / Conia-ene cyclization Aromatic nitration.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
168106-25-0 | |
625.651 | |
QEORIOGPVTWFMH-UHFFFAOYSA-N | |
131849436 | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2] |
C4F12N2O8S4Zn | |
MFCD16621474 | |
Zinc triflimide; Zinc trifluoromethanesulfonimide | |
zinc;bis(trifluoromethylsulfonyl)azanide |
Specifications
168106-25-0 | |
White | |
C4F12N2O8S4Zn | |
MFCD16621474 | |
Zinc triflimide; Zinc trifluoromethanesulfonimide | |
QEORIOGPVTWFMH-UHFFFAOYSA-N | |
zinc;bis(trifluoromethylsulfonyl)azanide | |
131849436 | |
Solid |
143°C to 147°C | |
>110°C (230°F) | |
[(CF3SO2)2N]2Zn | |
Moisture sensitive | |
It is soluble in CH2Cl2 and H2O. | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Zn+2] | |
625.651 | |
625.65 | |
Zinc bis (trifluoromethylsulfonyl)imide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
- Zinc bis(trifluoromethylsulfonyl)imide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Zinc bis(trifluoromethylsulfonyl)imide
RUO – Research Use Only