missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc bromide, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Mainly used in ocean wells as complete liquid, stationary liquid, amending liquid. It is also an important material in medical industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Specifications
Specifications
Boiling Point | 650°C |
Melting Point | 394°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
UN Number | UN3260 |
Merck Index | 14,10129 |
Solubility Information | Very soluble in ethanol and soluble in ethyl ether. Soluble in water (at 20°C 4470g/L). |
Formula Weight | 225.19 |
Percent Purity | 99.999% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'