missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc bromide, Puriss., anhydrous, ≥98%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
4 x 5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Physical Form | Anhydrous |
Quantity | 5 kg |
Linear Formula | ZnBr2 |
UN Number | 3260 |
Formula Weight | 225.2g/mol |
Percent Purity | ≥98% |
Grade | puriss. |
Chemical Name or Material | Zinc Bromide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'