missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc bromide, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Zinc bromide is an optimal catalyst for stereo specific and regioselective reaction of silacyclopropanes with carbonyl compounds. It is used as a transparent shield against radiation, as a fluid to displace drilling muds in transition phases, as a lewis acid in organic chemistry, colloidon emulsions for photography, a nutritional or functional ingredient in food and oral care products..
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
Boiling Point | 650°C |
Melting Point | 394°C |
Physical Form | Beads |
Quantity | 100 g |
UN Number | UN3260 |
Merck Index | 14,10129 |
Solubility Information | Soluble in water,ethanol,ether,90% alcohol. |
Formula Weight | 225.19 |
Percent Purity | 99.999% |
Packaging | Ampule |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'