missing translation for 'onlineSavingsMsg'
Learn More
Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals Product Code.: 11432008

Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals

Código de producto. 11432008
250 g
missing translation for 'orderingAttributeHoverText'
Quantity:
250 g
1000 g
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 11432008

Brand: Thermo Scientific Alfa Aesar A14590.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc carbonate basic is used as pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 5263-02-5
Molecular Formula C2H6O12Zn5
Molecular Weight (g/mol) 548.96
MDL Number MFCD00011519
InChI Key UOURRHZRLGCVDA-UHFFFAOYSA-D
Synonym Zinc subcarbonate; Zinc carbonate hydroxide
IUPAC Name pentazinc(2+) hexahydroxide dicarbonate
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O

Especificaciones

Quantity 250 g
Solubility Information Insoluble in water.
Formula Weight 548.97
Percent Purity 97%
Assay Zn >58.0%
Chemical Name or Material Zinc carbonate basic

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252191 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog NumberA14590.0B,A14590.30
Lot Number10248781 Certificate TypeCertificate of Analysis Date24/11/2023 Catalog NumberA14590.0B,A14590.30
Lot Number10246306 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA14590.30
Lot Number10246305 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA14590.30
Lot Number10237379 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA14590.0B,A14590.30
compliance-icons compliance-icons
Nombre del producto
  • Zinc carbonate, basic
Palabra de advertencia
  • Warning
Clasificación
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Indicaciones de peligro
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Otros peligros
  • MIXTURE LIST-Contains : Di[carbonato(2-)]hexahydroxypentazinc

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
Zinc carbonate basic, 97%, Zn >58.0%, Thermo Scientific Chemicals >

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado