missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 98%, extra pure

Product Code. 10531071 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
1 kg
2.5 kg
10 kg
Packaging:
Glass Bottle
Plastic drum
Unit Size:
10kg
1kg
2.5kg
5g
This item is not returnable. View return policy

Product Code. 10531071

Brand: Thermo Scientific Acros 196840010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7646-85-7 | Cl2Zn | 136.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.29
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976
IUPAC Name dichlorozinc
SMILES Cl[Zn]Cl

Specifications

Boiling Point 732.0°C
Melting Point 293.0°C
Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 1 kg
Assay Percent Range 98% min. (Complexometry)
Linear Formula ZnCl2
Fieser 01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391
Merck Index 15,10331
Solubility Information Solubility in water: 432 g/100mL (25°C). Other solubilities: soluble in alcohol, glycerol and ether, freely soluble in acetone
Formula Weight 136.29
Percent Purity ≥98%
Grade Extra Pure
Packaging Glass Bottle
Density .98g/mL
Chemical Name or Material Zinc chloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.