missing translation for 'onlineSavingsMsg'
Learn More

Zinc citrate dihydrate, Reagent Grade, Zn 31% min, Thermo Scientific Chemicals

Product Code. 11313417
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Zinc citrate dihydrate is used in dental care products such as toothpastes, mouthwashes and chewing gums due to its antimicrobial and anti-inflammatory effects. It used in used in food supplements, functional foods and beverages. It is used to formulated in pharmaceuticals, and foods as a zinc supplement. It used as intermediate in organic synthesis, agrochemicals and dyestuff. 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5990-32-9
Molecular Formula C12H14O16Zn3
Molecular Weight (g/mol) 610.368
MDL Number MFCD00150587
InChI Key OXAGUGIXGVHDGD-UHFFFAOYSA-H
Synonym zinc citrate dihydrate, unii-021ibv57il, acmc-20aj8r, citric acid zinc salt dihydrate, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt 2:3 , dihydrate, trizinc 2+ dicitrate dihydrate, zinc citrate, dihydrate, reagent, trizinc, ion zn2+ dicitrate dihydrate, zinc 2-hydroxypropane-1,2,3-tricarboxylate-water 3/2/2, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, zinc salt, hydrate 2:3:2
PubChem CID 16213234
IUPAC Name trizinc;2-hydroxypropane-1,2,3-tricarboxylate;dihydrate
SMILES C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.[Zn+2].[Zn+2].[Zn+2]

Specifications

Melting Point 334°C (decomposition)
Quantity 250 g
Solubility Information Soluble in dilute mineral acids and alkali hydroxides. Slightly soluble in water
Formula Weight 610.35 (574.32 Anhydrous)
Grade Reagent
Physical Form Powder
Chemical Name or Material Zinc citrate dihydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'