Learn More
Zinc fluoride, Puratronic™, 99.995% (metals basis), Thermo Scientific Chemicals
£55.70 - £584.00
Chemical Identifiers
CAS | 7783-49-5 |
---|---|
Molecular Formula | F2Zn |
Molecular Weight (g/mol) | 103.377 |
MDL Number | MFCD00011298 |
InChI Key | BHHYHSUAOQUXJK-UHFFFAOYSA-L |
Synonym | zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride |
PubChem CID | 10129912 |
IUPAC Name | zinc;difluoride |
SMILES | [F-].[F-].[Zn+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11302088
|
Thermo Scientific Alfa Aesar
012986.06 |
5 g |
£55.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11312088
|
Thermo Scientific Alfa Aesar
012986.14 |
25 g |
£178.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15466005
|
Thermo Scientific Alfa Aesar
012986.22 |
100 g |
£584.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is widely used in fluorination of organic compounds, as additive of electrolytic aluminum and also in electroplating baths. It is used in preserving food, manufacture of flux, oxygen-sensitive applications such as metal production, glasses, enamels, fabrics and plastics. It is also used in oil refining, manufacture of pharmaceuticals, manufacture phosphors for fluorescent electric lights, and floor coverings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7783-49-5 | |
103.377 | |
BHHYHSUAOQUXJK-UHFFFAOYSA-L | |
10129912 | |
[F-].[F-].[Zn+2] |
F2Zn | |
MFCD00011298 | |
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride | |
zinc;difluoride |
Specifications
1500°C | |
7783-49-5 | |
Powder | |
ZnF2 | |
14,10136 | |
Sparingly soluble in hydrochloric acid,nitric acid,and ammonium hydroxide. | |
[F-].[F-].[Zn+2] | |
103.377 | |
103.37 | |
Puratronic™ | |
(metals basis) |
872°C | |
White | |
F2Zn | |
MFCD00011298 | |
zinc ii fluoride, zinc 2+ ion difluoride, acmc-1bhy1, ksc379c0l, zinc fluoride 50g, zinc, ion zn2+ difluoride | |
BHHYHSUAOQUXJK-UHFFFAOYSA-L | |
zinc;difluoride | |
10129912 | |
99.995% | |
4.95 g/mL | |
Zinc fluoride |
Safety and Handling
GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P501c
H315-H318-H335
missing translation for 'einecsNumber' : 232-001-9
missing translation for 'rtecsNumber' : ZH3500000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only