missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc iodide, 98+%, pure, Thermo Scientific Chemicals
20.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 446°C |
Color | Beige-Yellow to White |
Physical Form | Hygroscopic Crystalline Powder or Crystals |
Quantity | 50 g |
Assay Percent Range | 98% min. (Argentometry) |
Linear Formula | ZnI2 |
Fieser | 01,1294; 05,765; 10,462; 11,604; 12,576; 13,88 |
Merck Index | 15, 10339 |
Solubility Information | Solubility in water: 4500g/L (20°C). Other solubilities: soluble in alkalies,soluble in alcohol and ether |
Formula Weight | 319.19 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'