missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Iodide-Starch Solution, Indicator, Reag. Ph. Eur., Honeywell™ Fluka™
Click to view available options
Quantity:
500 mL
Specifications
Quantity | 500 mL |
MDL Number | MFCD00148838 |
UN Number | UN3082 |
Chemical Name or Material | Zinc iodide-starch solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'