missing translation for 'onlineSavingsMsg'
Learn More

Zinc iron oxide

Product Code. 11383577
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11383577

Brand: Thermo Scientific Alfa Aesar 019407.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12063-19-3 | Fe2H2O4Zn | 243.082 g/mol

Zinc iron oxide is used as pigments, especially in applications related to heat stability. It is also used in high corrosion-resistant coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12063-19-3
Molecular Formula Fe2H2O4Zn
Molecular Weight (g/mol) 243.082
MDL Number MFCD00049621
InChI Key NNGHIEIYUJKFQS-UHFFFAOYSA-L
Synonym Zinc ferrite
PubChem CID 11831558
IUPAC Name hydroxy(oxo)iron;zinc
SMILES O[Fe]=O.O[Fe]=O.[Zn]

Specifications

Physical Form Fine Powder
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 241.06
Odor Odorless
Chemical Name or Material Zinc iron oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.