missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate, 6-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
655.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly Bottle
Specifications
Physical Form | Crystals |
Quantity | 2.5 kg |
Formula Weight | 297.47 |
Grade | BAKER ANALYZED™ Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 2.07kg |
Chemical Name or Material | Zinc Nitrate, 6-Hydrate |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS03_Oxidiser.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'