missing translation for 'onlineSavingsMsg'
Learn More

Zinc Nitrate Hexahydrate, 98%, Reagent grade, Honeywell™

Product Code. 15612280
Click to view available options
Quantity:
100 g
500 g
7 kg
Packaging:
Drum with inliner
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10196-18-6
Molecular Formula H12N2O12Zn
Molecular Weight (g/mol) 297.48
MDL Number MFCD00149889
InChI Key JGPSMWXKRPZZRG-UHFFFAOYSA-N
Synonym zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a
PubChem CID 15865313
SMILES O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 38.9°C
Color White
Quantity 100 g
Linear Formula Zn(NO3)2 · 6H2O
UN Number UN1514
Solubility Information Soluble in water
IUPAC Name zinc;dinitrate;hexahydrate
Formula Weight 297.49g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Powder or Crystals or Flakes
Density 2.060g/cm3
Chemical Name or Material Zinc Nitrate Hexahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'