missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate Hexahydrate, 98%, Reagent grade, Honeywell™
Click to view available options
Quantity:
100 g
500 g
7 kg
Packaging:
Drum with inliner
Poly Bottle
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 38.9°C |
Color | White |
Quantity | 7 kg |
Linear Formula | Zn(NO3)2 · 6H2O |
UN Number | UN1514 |
Solubility Information | Soluble in water |
IUPAC Name | zinc(2+) hexahydrate dinitrate |
Formula Weight | 297.49g/mol |
Percent Purity | 98% |
Grade | Reagent |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'