missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
Specifications
Specifications
Melting Point | 36°C |
Color | White |
pH | 5.1 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 297.46g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Zinc nitrate hexahydrate
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Oxidising solid Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Oxidising solid Category 2
- H272-May intensify fire; oxidiser.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zinc Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction