Learn More
Zinc nitrate hexahydrate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
£182.00 - £626.00
Chemical Identifiers
CAS | 10196-18-6 |
---|---|
Molecular Formula | H12N2O12Zn |
Molecular Weight (g/mol) | 297.48 |
MDL Number | MFCD00149889 |
InChI Key | JGPSMWXKRPZZRG-UHFFFAOYSA-N |
Synonym | zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a |
PubChem CID | 15865313 |
SMILES | O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316158
|
Thermo Scientific Alfa Aesar
011136.18 |
50 g |
£182.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326158
|
Thermo Scientific Alfa Aesar
011136.30 |
250 g |
£626.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zinc nitrate hexahydrate is used as an analytical reagent, catalyst and as a mordant in dyeing. It finds application in electro galvanizing, rubber industry and in the water treatment industry. It is widely used as the precursor for the synthesis of zinc oxide. It is employed in the preparation of the steel phosphating agent. In the field of agriculture, it is used for nourishing and protecting fruit trees from acid rain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10196-18-6 | |
297.48 | |
JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
15865313 |
H12N2O12Zn | |
MFCD00149889 | |
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a | |
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
∼36°C | |
White | |
H12N2O12Zn | |
MFCD00149889 | |
14,10144 | |
Soluble in water and alcohol. | |
O.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
297.48 | |
297.47 (189.38 Anhydrous) | |
Puratronic™ | |
Hygroscopic | |
(metals basis) |
10196-18-6 | |
Crystalline Aggregates | |
Zn(NO3)2·6H2O | |
UN1514 | |
zinc nitrate hexahydrate, zinc ii nitrte, zincnitratehexahydrate, acmc-1bzdd, n2o6zn.6h2o, ksc176g7t, zinc nitrate hexahydrate, puratronic, zinc nitrate, zinc 2+ hexahydrate dinitrate, zinc nitrate hexahydrate, p.a | |
JGPSMWXKRPZZRG-UHFFFAOYSA-N | |
zinc;dinitrate;hexahydrate | |
15865313 | |
99.998% | |
Odorless | |
2.07 g/mL | |
Zinc nitrate hexahydrate |
Safety and Handling
GHS H Statement
H272-H302-H315-H319-H335
May intensify fire
oxidizer.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P370+P378q-P501c
H272-H302-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: ZINC NITRATE
missing translation for 'einecsNumber' : 231-943-8
missing translation for 'rtecsNumber' : ZH4775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only