missing translation for 'onlineSavingsMsg'
Learn More

Zinc nitride, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 15412987
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Zinc nitride is useful in optical and electronic devices. It is doped with oxygen to prepare a potential nitride-based transparent conductor, which is used in semiconductors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-49-1
Molecular Formula N2Zn3
Molecular Weight (g/mol) 224.15
MDL Number MFCD00061327
InChI Key AKJVMGQSGCSQBU-UHFFFAOYSA-N
Synonym zinc nitride, zinc nitride zn3n2
PubChem CID 12130759
SMILES [Zn++].[N-]=[Zn].[N-]=[Zn]

Specifications

Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water. Soluble in hydrochloric acid.
IUPAC Name zinc(2+) bis(zincioazanide)
Formula Weight 224.12
Percent Purity 99%
Density 6.22 g/mL
Assay (metals basis)
Chemical Name or Material Zinc nitride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'