missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc oxide, ACS reagent, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
100 g
500 g
1 kg
5 kg
Packaging:
Glass Bottle
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 1975.0°C |
Color | White |
Physical Form | Powder |
Quantity | 5 g |
Linear Formula | ZnO |
Fieser | 01,1294; 05,622 |
Merck Index | 15,10347 |
Solubility Information | Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol |
Formula Weight | 81.38 |
Percent Purity | ≥99.0% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'