missing translation for 'onlineSavingsMsg'
Learn More

Zinc Oxide, Certified AR for Analysis, Fisher Chemical™

£117.00

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10294350
View Documents
Fisher Chemical
Z/1350/60
1 kg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

1314-13-2
81.38
XLOMVQKBTHCTTD-UHFFFAOYSA-N
14806
oxozinc
OZn
MFCD00011300
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
CHEBI:36560
O=[Zn]

Specifications

1975°C
White
Powder/Solid
MFCD00011300
XLOMVQKBTHCTTD-UHFFFAOYSA-N
oxozinc
14806
81.38g/mol
1314-13-2
7
OZn
zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
O=[Zn]
81.38
CHEBI:36560
Plastic powder jar
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Zinc oxide

Signal Word
  • Warning

Hazard Category
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P273-Avoid release to the environment.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.