missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc Oxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Poly Bottle
Specifications
Specifications
Physical Form | Powder |
Quantity | 500 g |
Formula Weight | 81.39 |
Grade | BAKER ANALYZED™ ACS Reagent |
Packaging | Poly Bottle |
Specific Gravity | 1L = 5.61kg |
Chemical Name or Material | Zinc Oxide |
Safety and Handling
Signal Word
- Warning
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'