missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 Ea.
Description
Zinc oxide is used as an additive in numerous materials and products including rubbers, plastics, ceramics, glass, cement, lubricants, paints, ointments, adhesives, sealants, pigments, foods, batteries, ferrites, fire retardants, and first-aid tapes. It is also used in oil, greases, ferrites, cosmetics, pharmaceuticals, medical, UV coatings, lacquers, ceramics and textiles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Quantity | 1 Ea. |
CAS | 1314-13-2 |
Molecular Formula | OZn |
MDL Number | MFCD00011300 |
Synonym | zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85 |
Solubility Information | Soluble in diluted acid. Insoluble in water and alcohol. |
InChI Key | XLOMVQKBTHCTTD-UHFFFAOYSA-N |
SMILES | O=[Zn] |
IUPAC Name | oxozinc |
Molecular Weight (g/mol) | 81.38 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'